Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gur-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062591.6 | NP_494992.2 | 7 - 8 | 1153 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |