Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062594.3 | NP_494995.2 | 12 - 13 | 4382 | 118 | |
NR_134556.1 | 12 - 13 | 4640 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |