Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062605.9 | NP_495006.2 | 13 - 14 | 2052 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |