Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062610.9 | NP_495011.1 | 5 - 6 | 715 | 72 | |
GenBank mRNA | AB007320.1 | - | 5 - 6 | 734 | 72 | |
AF025666.1 | - | 5 - 6 | 717 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |