Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062610.9 | NP_495011.1 | 6 - 7 | 971 | 87 | |
GenBank mRNA | AB007320.1 | - | 6 - 7 | 990 | 87 | |
AF025666.1 | - | 6 - 7 | 973 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |