Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062634.6 | NP_495035.1 | 2 - 3 | 404 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |