Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306583.2 | NP_001293512.1 | 2 - 3 | 217 | 91 | |
GenBank mRNA | AF015882.1 | - | 2 - 3 | 218 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |