Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025939.6 | NP_001021110.1 | 9 - 10 | 2582 | 74 | |
NM_001047257.6 | NP_001040722.1 | 1 - 2 | 71 | 74 | ||
NM_062788.7 | NP_495189.2 | 2 - 3 | 272 | 74 | ||
GenBank mRNA | AY091591.1 | - | 9 - 10 | 2543 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |