Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062856.7 | NP_495257.3 | 9 - 10 | 1709 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |