Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bpl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383861.1 | 11 - 12 | 1916 | 95 | ||
NM_062856.7 | NP_495257.3 | 11 - 12 | 1956 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |