Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062870.7 | NP_495271.1 | 1 - 2 | 169 | 90 | |
GenBank mRNA | AF104017.1 | - | 1 - 2 | 150 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |