Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062870.7 | NP_495271.1 | 7 - 8 | 850 | 86 | |
GenBank mRNA | AF104017.1 | - | 7 - 8 | 831 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |