Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062876.7 | NP_495277.1 | 1 - 2 | 320 | 77 | |
GenBank mRNA | AF326943.1 | - | 1 - 2 | 320 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |