Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001365320.2 | NP_001352251.1 | 8 - 9 | 610 | 117 | |
NM_001368591.4 | NP_001355419.1 | 7 | 1008 | 117 |
Gene Symbol: T25D10.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062891.5 | NP_495292.1 | 7 - 8 | 2620 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |