Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026958.5 | NP_001022129.1 | 1 - 2 | 126 | 129 | |
NM_062993.8 | NP_495394.3 | 1 - 2 | 121 | 129 | ||
GenBank mRNA | L15302.1 | - | 1 - 2 | 148 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |