Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyn-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026719.5 | NP_001021890.1 | 3 - 4 | 363 | 122 | |
NM_063015.5 | NP_495416.2 | 3 - 4 | 365 | 122 | ||
GenBank mRNA | U34954.1 | - | 3 - 4 | 362 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |