Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sqv-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063035.6 | NP_495436.1 | 4 - 5 | 547 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |