Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: him-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063050.6 | NP_495451.1 | 5 - 6 | 677 | 100 | |
GenBank mRNA | AF178755.1 | - | 5 - 6 | 677 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |