Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026889.5 | NP_001022060.1 | 15 - 16 | 2635 | 139 | |
NM_001313600.3 | NP_001300529.1 | 14 | 2523 | 139 | ||
NM_001313601.3 | NP_001300530.1 | 14 | 2496 | 139 | ||
NM_001313602.3 | NP_001300531.1 | 13 - 14 | 2406 | 139 | ||
GenBank mRNA | AF319614.1 | - | 12 - 13 | 2389 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |