Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026889.5 | NP_001022060.1 | 19 - 20 | 3261 | 86 | |
NM_001313600.3 | NP_001300529.1 | 18 | 3149 | 86 | ||
NM_001313601.3 | NP_001300530.1 | 18 | 3122 | 86 | ||
NM_001313602.3 | NP_001300531.1 | 17 - 18 | 3032 | 86 | ||
GenBank mRNA | AF319614.1 | - | 16 - 17 | 3015 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |