Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063124.7 | NP_495525.2 | 5 - 6 | 451 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |