Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026929.4 | NP_001022100.1 | 9 - 10 | 1258 | 120 | |
NM_001026930.4 | NP_001022101.1 | 8 - 9 | 1779 | 120 | ||
NM_001026931.6 | NP_001022102.1 | 9 - 10 | 1267 | 120 | ||
NM_001026932.5 | NP_001022103.1 | 9 - 10 | 1225 | 120 | ||
NM_001047301.4 | NP_001040766.1 | 9 - 10 | 1271 | 120 | ||
GenBank mRNA | AF083234.1 | - | 7 - 8 | 1156 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |