Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026930.4 | NP_001022101.1 | 2 - 3 | 780 | 67 | |
GenBank mRNA | AF083234.1 | - | 1 - 2 | 157 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |