Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: insc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381495.1 | NP_001368642.1 | 11 - 12 | 1238 | 114 | |
NM_001381496.2 | NP_001367460.1 | 11 - 12 | 1262 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |