Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bath-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063148.6 | NP_495549.1 | 8 - 9 | 1152 | 67 | |
NM_063149.5 | NP_495550.1 | 8 - 9 | 1131 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |