Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351919.3 | NP_001338828.1 | 12 - 13 | 2421 | 118 | |
NM_001351920.3 | NP_001338797.1 | 11 - 12 | 2408 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |