Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351919.3 | NP_001338828.1 | 14 - 15 | 2965 | 86 | |
NM_001351920.3 | NP_001338797.1 | 13 - 14 | 2952 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |