Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351919.3 | NP_001338828.1 | 15 - 16 | 3159 | 147 | |
NM_001351920.3 | NP_001338797.1 | 14 - 15 | 3146 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |