Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sro-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377819.1 | NP_001364737.1 | 3 - 4 | 260 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |