Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aka-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026874.4 | NP_001022045.1 | 12 - 13 | 3158 | 130 | |
NM_001383888.1 | 13 - 14 | 3059 | 130 | |||
NM_063164.8 | NP_495565.2 | 12 - 13 | 3170 | 130 | ||
GenBank mRNA | AF045926.1 | - | 12 - 13 | 3169 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |