Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wrt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063178.8 | NP_495579.3 | 1 - 2 | 387 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |