Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026714.4 | NP_001021885.1 | 2 - 3 | 304 | 99 | |
NM_001026715.6 | NP_001021886.1 | 5 - 6 | 1772 | 99 | ||
GenBank mRNA | DQ241299.1 | - | 2 - 3 | 304 | 99 | |
DQ241300.1 | - | 3 - 4 | 345 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |