Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cki-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027138.6 | NP_001022309.1 | 5 - 6 | 779 | 106 | |
NM_063241.7 | NP_495642.3 | 5 - 6 | 777 | 106 | ||
GenBank mRNA | AF179359.1 | - | 4 - 5 | 673 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |