Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027161.3 | NP_001022332.1 | 5 - 6 | 1076 | 70 | |
NM_001377823.1 | NP_001364642.1 | 5 - 6 | 1049 | 70 | ||
NM_001377824.1 | NP_001364641.1 | 5 - 6 | 1025 | 70 | ||
NM_001377825.1 | NP_001364552.1 | 5 - 6 | 998 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |