Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ufd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027149.5 | NP_001022320.1 | 5 - 6 | 2568 | 124 | |
NM_001383900.1 | 5 - 6 | 2569 | 124 | |||
NM_063290.3 | NP_495691.1 | 5 - 6 | 2583 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |