Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ltd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322746.3 | NP_001309573.1 | 3 - 4 | 543 | 81 | |
NM_063296.6 | NP_495697.2 | 6 - 7 | 1310 | 81 | ||
GenBank mRNA | AF489839.1 | - | 6 - 7 | 1332 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |