Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063312.7 | NP_495713.2 | 11 - 12 | 4431 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |