Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027145.3 | NP_001022316.1 | 1 - 2 | 137 | 64 | |
NM_001027146.3 | NP_001022317.1 | 1 - 2 | 49 | 64 | ||
NM_001027147.5 | NP_001022318.1 | 1 - 2 | 51 | 64 | ||
GenBank mRNA | AF063244.1 | - | 1 - 2 | 71 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |