Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027088.7 | NP_001022259.1 | 8 - 9 | 3228 | 84 | |
NM_001027089.7 | NP_001022260.1 | 7 - 8 | 3096 | 84 | ||
NM_001373755.2 | NP_001359723.1 | 7 - 8 | 3115 | 84 | ||
NM_001381521.1 | NP_001368728.1 | 7 - 8 | 3089 | 84 | ||
NM_001383902.1 | 8 - 9 | 3189 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |