Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tdc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063342.6 | NP_495743.1 | 6 - 7 | 1465 | 84 | |
NM_063343.4 | NP_495744.1 | 6 - 7 | 1475 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |