Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zyg-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063383.6 | NP_495784.1 | 5 - 6 | 1871 | 93 | |
GenBank mRNA | AF035197.1 | - | 5 - 6 | 1871 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |