Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313555.3 | NP_001300484.1 | 4 - 5 | 507 | 96 | |
NM_001313670.4 | NP_001300599.1 | 3 - 4 | 414 | 96 | ||
NM_063405.4 | NP_495806.1 | 2 - 3 | 308 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |