Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026848.5 | NP_001022019.1 | 3 - 4 | 635 | 119 | |
NM_001026849.3 | NP_001022020.1 | 3 - 4 | 631 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |