Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026848.5 | NP_001022019.1 | 4 - 5 | 864 | 63 | |
NM_001026849.3 | NP_001022020.1 | 4 - 5 | 860 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |