Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063450.3 | NP_495851.2 | 3 - 4 | 585 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |