Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330955.2 | NP_001317778.1 | 5 - 6 | 723 | 80 | |
NM_001381544.1 | NP_001368599.1 | 5 - 6 | 687 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |