Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063557.6 | NP_495958.1 | 10 - 11 | 2246 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |