Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sym-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063559.9 | NP_495960.2 | 9 - 10 | 1657 | 82 | |
GenBank mRNA | AY220985.1 | - | 3 - 4 | 661 | 82 |
Gene Symbol: twk-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356885.2 | NP_001343566.1 | 1964 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |