Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: let-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136359.5 | NP_001129831.1 | 2 - 3 | 197 | 155 | |
NM_001313678.3 | NP_001300607.1 | 3 - 4 | 224 | 155 | ||
NM_063561.5 | NP_495962.2 | 2 - 3 | 271 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |