Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mel-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026757.4 | NP_001021928.1 | 12 - 13 | 2829 | 115 | |
NM_001381562.1 | NP_001367038.1 | 11 - 12 | 2519 | 115 | ||
NM_001383917.1 | 10 - 11 | 2366 | 115 | |||
NM_063593.6 | NP_495994.1 | 11 - 12 | 2511 | 115 | ||
GenBank mRNA | U86640.1 | - | 11 - 12 | 2502 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |